Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr. 19: 53882184 - 53907647 on Build GRCh38
UniGene ID
Hs.631564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316329.1 NP_001303258.1 6 858
NM_002739.4 NP_002730.1 6 858
GenBank mRNA AB209671.1 - 6 577
AF345987.1 - 6 742
AK296412.1 - 6 866
AK296434.1 - 6 845
AK302958.1 - 6 843
AK303741.1 - 4 265
AK312302.1 - 6 820
BC047876.1 - 6 733 5764695
BI757265.1 - 6 841
M13977.1 - 6 742
Z15114.1 - 2 66

View Full Product Details