Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr. 19: 53882184 - 53907647 on Build GRCh38
UniGene ID
Hs.631564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316329.1 NP_001303258.1 7, 8 1143
NM_002739.4 NP_002730.1 7, 8 1143
GenBank mRNA AB209671.1 - 6, 7 1540
AF345987.1 - 7, 8 1027
AK296412.1 - 7, 8 1151
AK296434.1 - 7, 8 1130
AK303741.1 - 5, 6 550
AK312302.1 - 7, 8 1105
BC047876.1 - 7, 8 1018 5764695
M13977.1 - 7, 8 1027
Z15114.1 - 3, 4 351

View Full Product Details