Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242413.2 NP_001229342.1 11 1154
NM_001282644.1 NP_001269573.1 11 1152
NM_001282645.1 NP_001269574.1 10 1093
NM_001323265.1 NP_001310194.1 11 1209
NM_001323266.1 NP_001310195.1 10 1091
NM_001323267.1 NP_001310196.1 11 1152
NM_006257.4 NP_006248.1 11 1154
XM_005252496.4 XP_005252553.1 11 1254
XM_005252497.4 XP_005252554.1 11 1256
XM_006717465.3 XP_006717528.1 13 3649
XM_017016410.1 XP_016871899.1 12 3560
XM_017016411.1 XP_016871900.1 12 3588
GenBank mRNA AK293935.1 - 10 1031
AK308099.1 - 11 1127
AK312411.1 - 11 1104
AY702977.1 - 10 1054
BC101465.1 - 10 1073 8068971
BC113359.1 - 10 1073 8322411
BX647657.1 - 10 1239
L01087.1 - 11 1148
L07032.1 - 10 1075

View Full Product Details