Gene Transcripts
- Gene Symbol
- Olfr325
- Entrez Gene ID
- 258261
- Gene Name
- olfactory receptor 325
- Gene Aliases
- MOR275-5
- Chromosome Location
- Chr. 11: 58580837 - 58581877 on Build GRCm38
- UniGene ID
- Mm.303171
- Species
- Mus musculus
- Gene Symbol
- Olfr224
- Entrez Gene ID
- 258198
- Gene Name
- olfactory receptor 224
- Gene Aliases
- MOR275-3, Olfr224-ps1, Olfr327-ps1
- Chromosome Location
- Chr. 11: 58566399 - 58567343 on Build GRCm38
- UniGene ID
- Mm.480307
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207695.1 |
NP_997578.1
|
1 |
79 |
|
- Gene Symbol
- Olfr328
- Entrez Gene ID
- 258495
- Gene Name
- olfactory receptor 328
- Gene Aliases
- MOR275-2
- Chromosome Location
- Chr. 11: 58551305 - 58552237 on Build GRCm38
- UniGene ID
- Mm.339498
- Species
- Mus musculus
- Gene Symbol
- Olfr329-ps
- Entrez Gene ID
- 259148
- Gene Name
- olfactory receptor 329, pseudogene
- Gene Aliases
- MOR275-6P, Olfr329
- Chromosome Location
- Chr. 11: 58542447 - 58552797 on Build GRCm38
- UniGene ID
- Mm.339495
- Species
- Mus musculus
- Gene Symbol
- Olfr330
- Entrez Gene ID
- 258879
- Gene Name
- olfactory receptor 330
- Gene Aliases
- MOR275-1
- Chromosome Location
- Chr. 11: 58528929 - 58534825 on Build GRCm38
- UniGene ID
- Mm.223292
- Species
- Mus musculus
View Full Product Details