Gene Transcripts

Gene Symbol
Olfr913
Entrez Gene ID
258225
Gene Name
olfactory receptor 913
Gene Aliases
MOR165-10, MOR165-9P
Chromosome Location
Chr. 9: 38592803 - 38595161 on Build GRCm38
UniGene ID
Mm.335071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011523.2 NP_001011523.2 2 1135

View Full Product Details