Gene Transcripts

Gene Symbol
Olfr448
Entrez Gene ID
258270
Gene Name
olfactory receptor 448
Gene Aliases
MOR261-13
Chromosome Location
Chr. 6: 42896453 - 42897385 on Build GRCm38
UniGene ID
Mm.377404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146273.1 NP_666385.1 1 582
GenBank mRNA BC107336.1 - 1 574
BC107337.1 - 1 582 40055003

View Full Product Details