Gene Transcripts

Gene Symbol
Olfr187
Entrez Gene ID
258319
Gene Name
olfactory receptor 187
Gene Aliases
MOR183-8
Chromosome Location
Chr. 16: 59035780 - 59039749 on Build GRCm38
UniGene ID
Mm.223015
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146322.2 NP_666434.2 2 757
GenBank mRNA BC119304.1 - 1 645
BC119306.1 - 1 648 8734055

View Full Product Details