Gene Transcripts
- Gene Symbol
- Olfr731
- Entrez Gene ID
- 258360
- Gene Name
- olfactory receptor 731
- Gene Aliases
- MOR246-5
- Chromosome Location
- Chr. 14: 50237903 - 50238883 on Build GRCm38
- UniGene ID
- Mm.377457
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146363.2 |
NP_666475.2
|
1 |
475 |
|
View Full Product Details