Gene Transcripts

Gene Symbol
Olfr1368
Entrez Gene ID
258527
Gene Name
olfactory receptor 1368
Gene Aliases
MOR256-15
Chromosome Location
Chr. 13: 21142111 - 21143055 on Build GRCm38
UniGene ID
Mm.271440
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146534.1 NP_666745.1 1 706
GenBank mRNA BC125382.1 - 1 732
BC125384.1 - 1 732 40129899

View Full Product Details