Gene Transcripts

Gene Symbol
Olfr912
Entrez Gene ID
258806
Gene Name
olfactory receptor 912
Gene Aliases
MOR165-4
Chromosome Location
Chr. 9: 38580244 - 38582211 on Build GRCm38
UniGene ID
Mm.223233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146810.2 NP_667021.2 2 725
GenBank mRNA CB174340.1 - 1 399

View Full Product Details