Gene Transcripts
- Gene Symbol
- Olfr912
- Entrez Gene ID
- 258806
- Gene Name
- olfactory receptor 912
- Gene Aliases
- MOR165-4
- Chromosome Location
- Chr. 9: 38580244 - 38582211 on Build GRCm38
- UniGene ID
- Mm.223233
- Species
- Mus musculus
- Gene Symbol
- Olfr910
- Entrez Gene ID
- 258807
- Gene Name
- olfactory receptor 910
- Gene Aliases
- MOR165-3
- Chromosome Location
- Chr. 9: 38537882 - 38539829 on Build GRCm38
- UniGene ID
- Mm.223234
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146811.2 |
NP_667022.1
|
2 |
140 |
|
View Full Product Details