Gene Transcripts

Gene Symbol
PSG2
Entrez Gene ID
5670
Gene Name
pregnancy specific beta-1-glycoprotein 2
Gene Aliases
CEA, PSBG2, PSG1
Chromosome Location
Chr. 19: 43064210 - 43082741 on Build GRCh38
UniGene ID
Hs.709200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031246.3 NP_112536.2 4 1118
GenBank mRNA M20882.1 - 4 1029
M37398.1 - 1 219

View Full Product Details