Gene Transcripts
- Gene Symbol
- Olfr545
- Entrez Gene ID
- 258837
- Gene Name
- olfactory receptor 545
- Gene Aliases
- MOR42-1, S50
- Chromosome Location
- Chr. 7: 102493823 - 102494773 on Build GRCm38
- UniGene ID
- Mm.377714
- Species
- Mus musculus
- Gene Symbol
- Olfr544
- Entrez Gene ID
- 257926
- Gene Name
- olfactory receptor 544
- Gene Aliases
- MOR42-3, Ors6
- Chromosome Location
- Chr. 7: 102484114 - 102488307 on Build GRCm38
- UniGene ID
- Mm.103733
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020289.2 |
NP_064685.2
|
2 |
479 |
|
View Full Product Details