Gene Transcripts

Gene Symbol
Olfr275
Entrez Gene ID
258857
Gene Name
olfactory receptor 275
Gene Aliases
MOR262-2
Chromosome Location
Chr. 4: 52825399 - 52826358 on Build GRCm38
UniGene ID
Mm.484340
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146858.2 NP_667069.2 1 73
GenBank mRNA BC101951.1 - 1 72
BC141896.1 - 1 65

View Full Product Details