Gene Transcripts
- Gene Symbol
- PSMA7
- Entrez Gene ID
- 5688
- Gene Name
- proteasome subunit alpha 7
- Gene Aliases
- C6, HEL-S-276, HSPC, RC6-1, XAPC7
- Chromosome Location
- Chr. 20: 62136727 - 62143458 on Build GRCh38
- UniGene ID
- Hs.233952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002792.3 |
NP_002783.1
|
6, 7 |
806 |
|
GenBank mRNA |
AF022815.1 |
- |
6, 7 |
675 |
|
AF054185.1 |
- |
6, 7 |
736 |
|
AK312025.1 |
- |
6, 7 |
701 |
|
BC001895.1 |
- |
6, 7 |
739 |
|
BC004427.1 |
- |
6, 7 |
724 |
2819923 |
BC014999.1 |
- |
6, 7 |
734 |
|
BG575009.1 |
- |
6, 7 |
800 |
|
BT007165.1 |
- |
6, 7 |
651 |
|
GQ472224.1 |
- |
6, 7 |
724 |
|
View Full Product Details