Gene Transcripts
- Gene Symbol
- Olfr381
- Entrez Gene ID
- 259024
- Gene Name
- olfactory receptor 381
- Gene Aliases
- MOR135-4
- Chromosome Location
- Chr. 11: 73485887 - 73486822 on Build GRCm38
- UniGene ID
- Mm.390415
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_147022.2 |
NP_667233.2
|
1 |
454 |
|
View Full Product Details