Gene Transcripts

Gene Symbol
PSMB5
Entrez Gene ID
5693
Gene Name
proteasome subunit beta 5
Gene Aliases
LMPX, MB1, X
Chromosome Location
Chr. 14: 23016543 - 23035220 on Build GRCh38
UniGene ID
Hs.422990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130725.1 NP_001124197.1 2 128
NM_001144932.2 NP_001138404.1 2 481
NM_002797.4 NP_002788.1 2 481
XM_005267871.2 XP_005267928.1 2 476
GenBank mRNA AK300714.1 - 2 226
AK311895.1 - 2 237
BC057840.1 - 2 236 4795732
BC107720.1 - 2 232 6738409
BT006777.1 - 2 52
BU539357.1 - 2 104
BX248299.1 - 2 213
BX538001.1 - 2 228
CD048996.1 - 2 160
S74378.1 - 2 73

View Full Product Details