Gene Transcripts

Gene Symbol
Olfr582
Entrez Gene ID
259055
Gene Name
olfactory receptor 582
Gene Aliases
MOR30-3
Chromosome Location
Chr. 7: 103041481 - 103042440 on Build GRCm38
UniGene ID
Mm.377812
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147053.1 NP_667264.1 1 664
GenBank mRNA BC107338.1 - 1 664
BC107339.1 - 1 664

View Full Product Details