Gene Transcripts

Gene Symbol
Plekhg3
Entrez Gene ID
263406
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 3
Gene Aliases
BC030417
Chromosome Location
Chr. 12: 76533247 - 76579042 on Build GRCm38
UniGene ID
Mm.291854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153804.4 NP_722499.4 2, 3 658
GenBank mRNA AK080928.1 - 2, 3 659
AK162025.1 - 2, 3 600
BC096443.1 - 2, 3 636 4189193

View Full Product Details