Gene Transcripts

Gene Symbol
Xxylt1
Entrez Gene ID
268880
Gene Name
xyloside xylosyltransferase 1
Gene Aliases
AI480653
Chromosome Location
Chr. 16: 30928348 - 31081551 on Build GRCm38
UniGene ID
Mm.286693
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198626.2 NP_941028.2 4 977
GenBank mRNA AK154256.1 - 4 977
AK166259.1 - 4 930
BC031419.1 - 4 925 2615706

View Full Product Details