Gene Transcripts

Gene Symbol
Wdr3
Entrez Gene ID
269470
Gene Name
WD repeat domain 3
Gene Aliases
AW546279, D030020G18Rik
Chromosome Location
Chr. 3: 100138180 - 100162403 on Build GRCm38
UniGene ID
Mm.24591
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175552.4 NP_780761.1 7, 8 891
GenBank mRNA AK043557.1 - 7, 8 824
AK050738.1 - 7, 8 859
AK050797.1 - 7, 8 862
BC063100.1 - 7, 8 877 6332749

View Full Product Details