Gene Transcripts

Gene Symbol
Chsy1
Entrez Gene ID
269941
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
mKIAA0990
Chromosome Location
Chr. 7: 66109515 - 66173798 on Build GRCm38
UniGene ID
Mm.333916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081163.1 NP_001074632.1 3 2164
GenBank mRNA AK129255.1 - 3 2164
BC128567.1 - 4 1611
BC153826.1 - 2 702
BC153827.1 - 4 1636

View Full Product Details