Gene Transcripts

Gene Symbol
Phf21b
Entrez Gene ID
271305
Gene Name
PHD finger protein 21B
Gene Aliases
A730032D07Rik
Chromosome Location
Chr. 15: 84785376 - 84856132 on Build GRCm38
UniGene ID
Mm.101022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081166.2 NP_001074635.2 13 1948
GenBank mRNA AK033659.1 - 2 401
AK042863.1 - 13 1539
AK046530.1 - 11 2203
BC044847.1 - 1 2800
BC048859.1 - 1 2800
BC059040.1 - 1 768
BC067021.1 - 13 1592
BC157925.1 - 13 1570 9088098
BC158044.1 - 13 1570 9007348
BC172019.1 - 13 1570 9054305
BC172145.1 - 12 1547 9054815

View Full Product Details