Gene Transcripts

Gene Symbol
Cdk15
Entrez Gene ID
271697
Gene Name
cyclin-dependent kinase 15
Gene Aliases
Als2cr7, Pftk2
Chromosome Location
Chr. 1: 59256889 - 59352612 on Build GRCm38
UniGene ID
Mm.297713
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033373.2 NP_001028545.2 9 927
GenBank mRNA AK042881.1 - 2 136
AK211120.1 - 2 72
BC147314.1 - 9 927 9056364
BC147315.1 - 9 927 9056370
BC147758.1 - 9 927 9087483
BC147763.1 - 9 927 9087488
BC172036.1 - 9 924 9054322

View Full Product Details