Gene Transcripts

Gene Symbol
BC089491
Entrez Gene ID
280621
Gene Name
cDNA sequence BC089491
Gene Aliases
Selv
Chromosome Location
Chr. 7: 28284652 - 28291134 on Build GRCm38
UniGene ID
Mm.212777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175033.3 NP_778198.2 1 736
GenBank mRNA AK077113.2 - 1 789
BC089491.1 - 1 735 6772991

View Full Product Details