Gene Transcripts
- Gene Symbol
- Hist1h2ac
- Entrez Gene ID
- 319164
- Gene Name
- histone cluster 1, H2ac
- Gene Aliases
- Chromosome Location
- Chr. 13: 23683473 - 23683959 on Build GRCm38
- UniGene ID
- Mm.261973
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033518.1 |
- |
|
609 |
|
View Full Product Details