Gene Transcripts

Gene Symbol
C5ar2
Entrez Gene ID
319430
Gene Name
complement component 5a receptor 2
Gene Aliases
C5L2, E030029A11Rik, Gpr77
Chromosome Location
Chr. 7: 16234585 - 16244154 on Build GRCm38
UniGene ID
Mm.239570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146005.1 NP_001139477.1 2 683
NM_176912.4 NP_795886.2 2 776
GenBank mRNA AK053187.1 - 2 777
AK160050.1 - 2 684
BC137941.1 - 2 665 8860960
BC145834.1 - 2 665 40131137

View Full Product Details