Gene Transcripts

Gene Symbol
Ccdc66
Entrez Gene ID
320234
Gene Name
coiled-coil domain containing 66
Gene Aliases
E230015L20Rik
Chromosome Location
Chr. 14: 27481090 - 27508506 on Build GRCm38
UniGene ID
Mm.247767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177111.3 NP_796085.3 4 374
GenBank mRNA AK145297.1 - 3 387
AK170006.1 - 4 422
BC070471.1 - 4 374 30626499

View Full Product Details