Gene Transcripts

Gene Symbol
Tnfsf15
Entrez Gene ID
326623
Gene Name
tumor necrosis factor (ligand) superfamily, member 15
Gene Aliases
Tl1, Tl1a, Vegi
Chromosome Location
Chr. 4: 63724603 - 63745113 on Build GRCm38
UniGene ID
Mm.208152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177371.3 NP_796345.3 1 292
GenBank mRNA AF520786.1 - 1 209
AY764130.1 - 1 292
BC096494.1 - 1 278 30740802
BC138207.1 - 1 257 8861226
BC138208.1 - 1 257 8861227

View Full Product Details