Gene Transcripts

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
Chromosome Location
Chr. 21: 44107262 - 44131181 on Build GRCh38
UniGene ID
Hs.449076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005049.2 NP_005040.2 16 2072
XM_011529667.2 XP_011527969.1 16 2137
GenBank mRNA AB209438.1 - 5 3516
AK314184.1 - 16 2076
BC013309.2 - 16 1982 3528790
BC014988.1 - 16 1982 4906611
BX387451.2 - 4 382
U53346.1 - 16 2006
U56085.1 - 16 2017
X95263.1 - 16 2013

Gene Symbol
LOC102724159
Entrez Gene ID
102724159
Gene Name
periodic tryptophan protein 2 homolog
Gene Aliases
Chromosome Location
Chr. 21: 5130871 - 5154767 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006723910.1 XP_006723973.1 16 2109

View Full Product Details