Gene Transcripts

Gene Symbol
Slc39a9
Entrez Gene ID
328133
Gene Name
solute carrier family 39 (zinc transporter), member 9
Gene Aliases
2010002A02Rik, 2610511I23Rik, 4833420E20Rik, AW209151
Chromosome Location
Chr. 12: 80644215 - 80683342 on Build GRCm38
UniGene ID
Mm.238279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026244.2 NP_080520.2 6, 7 1359
GenBank mRNA AK080462.1 - 6, 7 1341
AK090062.1 - 6, 7 1352
BC043112.1 - 6, 7 1306
BC072655.1 - 6, 7 1335
BC117754.1 - 6, 7 953 40090588
BC117755.1 - 6, 7 953 40090589
BC158056.1 - 6, 7 1104 9088164

View Full Product Details