Gene Transcripts

Gene Symbol
Parp4
Entrez Gene ID
328417
Gene Name
poly (ADP-ribose) polymerase family, member 4
Gene Aliases
Adprtl1, C030027K23Rik, E230037B21Rik, Gm743, PARPL, PH5P, VAULT3, VPARP, p193
Chromosome Location
Chr. 14: 56575619 - 56659798 on Build GRCm38
UniGene ID
Mm.379357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145978.2 NP_001139450.2 8, 9 973
GenBank mRNA AK054229.1 - 8, 9 937
AK164513.1 - 8, 9 973
AK172907.1 - 8, 9 905

View Full Product Details