Gene Transcripts

Gene Symbol
Nostrin
Entrez Gene ID
329416
Gene Name
nitric oxide synthase trafficker
Gene Aliases
Daip2, mDaIP2
Chromosome Location
Chr. 2: 69135800 - 69189330 on Build GRCm38
UniGene ID
Mm.90047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181547.3 NP_853525.3 6 463
GenBank mRNA AJ564264.1 - 6 373
AK052926.1 - 6 455
AY280963.1 - 6 416
BC069942.1 - 6 440 30443942

View Full Product Details