Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 774 | |
XM_005266733.1 | XP_005266790.1 | 7 | 695 | ||
XM_017025913.1 | XP_016881402.1 | 9 | 1528 | ||
XM_017025914.1 | XP_016881403.1 | 8 | 923 | ||
GenBank mRNA | AF329499.1 | - | 6 | 774 | |
AF498954.1 | - | 5 | 531 | ||
AK314115.1 | - | 6 | 1052 | ||
BC027474.1 | - | 6 | 642 | 4520182 | |
U57093.2 | - | 6 | 623 |