Gene Transcripts

Gene Symbol
RAB5C
Entrez Gene ID
5878
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
L1880, RAB5CL, RAB5L, RABL
Chromosome Location
Chr. 17: 42124976 - 42155044 on Build GRCh38
UniGene ID
Hs.650382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252039.1 NP_001238968.1 6 961
NM_004583.3 NP_004574.2 5 745
NM_201434.2 NP_958842.1 6 862
XM_011525090.1 XP_011523392.1 5 684
XM_011525091.1 XP_011523393.1 5 808
GenBank mRNA AF141304.1 - 4 574
AF498938.1 - 4 505
AK310234.1 - 6 905
AL553867.3 - 5 700
BC106039.1 - 5 704 4938169
BC114439.1 - 5 700 40083572
BM424166.1 - 6 802
BM550607.1 - 5 705
BT019484.1 - 4 505
CR541901.1 - 4 505
U11293.1 - 4 580
U18420.1 - 5 640

View Full Product Details