Gene Transcripts

Gene Symbol
Sec14l3
Entrez Gene ID
380683
Gene Name
SEC14-like 3 (S. cerevisiae)
Gene Aliases
1110069O07Rik
Chromosome Location
Chr. 11: 4064853 - 4078990 on Build GRCm38
UniGene ID
Mm.296806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029937.2 NP_001025108.1 9 786
GenBank mRNA AK136060.1 - 3 139
BC138982.1 - 9 786 8862004
BC138985.1 - 9 786 8862007
BC145485.1 - 9 829

View Full Product Details