Gene Transcripts

Gene Symbol
Oog2
Entrez Gene ID
381570
Gene Name
oogenesin 2
Gene Aliases
C85069
Chromosome Location
Chr. 4: 144190717 - 144196934 on Build GRCm38
UniGene ID
Mm.332028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198661.3 NP_941063.1 5 1405
GenBank mRNA AK139660.1 - 3 2607
AY573562.1 - 5 1405
BC052839.1 - 5 1405 30050073

View Full Product Details