Gene Transcripts

Gene Symbol
Fbxw22
Entrez Gene ID
382156
Gene Name
F-box and WD-40 domain protein 22
Gene Aliases
EG382156, Gm5164
Chromosome Location
Chr. 9: 109378400 - 109404294 on Build GRCm38
UniGene ID
Mm.373875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014395.2 NP_001014417.1 9 1301
GenBank mRNA AK135808.1 - 7 1018
AK162068.1 - 5 809
BC084593.1 - 9 1305 30477093

View Full Product Details