Gene Transcripts

Gene Symbol
Taar8b
Entrez Gene ID
382348
Gene Name
trace amine-associated receptor 8B
Gene Aliases
Gm1149, taR-8b
Chromosome Location
Chr. 10: 24091260 - 24092294 on Build GRCm38
UniGene ID
Mm.377897
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010837.1 NP_001010837.1 1 700
GenBank mRNA BC139105.1 - 1 726
BC139106.1 - 1 726 8862128

View Full Product Details