Gene Transcripts

Gene Symbol
I830077J02Rik
Entrez Gene ID
433638
Gene Name
RIKEN cDNA I830077J02 gene
Gene Aliases
Chromosome Location
Chr. 3: 105925851 - 105932931 on Build GRCm38
UniGene ID
Mm.239081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033780.3 NP_001028952.1 6 432
GenBank mRNA AK152510.1 - 6 435
BC147821.1 - 6 372 9087538
BC147828.1 - 6 372 9087545

View Full Product Details