Gene Transcripts
- Gene Symbol
- ROBO1
- Entrez Gene ID
- 6091
- Gene Name
- roundabout guidance receptor 1
- Gene Aliases
- DUTT1, SAX3
- Chromosome Location
- Chr. 3: 78597238 - 79767998 on Build GRCh38
- UniGene ID
- Hs.744218
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002941.3 |
NP_002932.1
|
20, 21 |
3044 |
|
XM_006713277.2 |
XP_006713340.1
|
19, 20 |
3695 |
|
XM_011533976.1 |
XP_011532278.1
|
21, 22 |
3142 |
|
XM_011533977.2 |
XP_011532279.1
|
21, 22 |
2995 |
|
XM_011533979.1 |
XP_011532281.1
|
21, 22 |
3142 |
|
XM_017006982.1 |
XP_016862471.1
|
20, 21 |
3054 |
|
XM_017006983.1 |
XP_016862472.1
|
18, 19 |
3686 |
|
XM_017006984.1 |
XP_016862473.1
|
20, 21 |
3133 |
|
XM_017006985.1 |
XP_016862474.1
|
19, 20 |
2795 |
|
GenBank mRNA |
AF040990.1 |
- |
19, 20 |
2836 |
|
AK299920.1 |
- |
10, 11 |
1733 |
|
BX538319.1 |
- |
12, 13 |
1755 |
|
View Full Product Details