Gene Transcripts

Gene Symbol
Xlr5a
Entrez Gene ID
574438
Gene Name
X-linked lymphocyte-regulated 5A
Gene Aliases
Xlr5d
Chromosome Location
Chr. X: 73107621 - 73119524 on Build GRCm38
UniGene ID
Mm.435653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045539.2 NP_001039004.2 9 790
GenBank mRNA BC051681.1 - 9 790

View Full Product Details