Gene Transcripts

Gene Symbol
Kctd21
Entrez Gene ID
622320
Gene Name
potassium channel tetramerisation domain containing 21
Gene Aliases
EG622320
Chromosome Location
Chr. 7: 97332311 - 97350216 on Build GRCm38
UniGene ID
Mm.334029
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039039.3 NP_001034128.2 2 223
GenBank mRNA AK141547.1 - 2 220
BB852194.1 - 2 224
BC139363.1 - 1 172 8862385
BC139365.1 - 1 172 8862387

View Full Product Details