Gene Transcripts
- Gene Symbol
- 1700110I01Rik
- Entrez Gene ID
- 73549
- Gene Name
- RIKEN cDNA 1700110I01 gene
- Gene Aliases
- 100040740, 1700016F01Rik, Gm2938
- Chromosome Location
- Chr. 14: 3262402 - 3305831 on Build GRCm38
- UniGene ID
- Mm.398741
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007159.1 |
- |
4 |
1013 |
|
View Full Product Details