Gene Transcripts

Gene Symbol
Lama2
Entrez Gene ID
16773
Gene Name
laminin, alpha 2
Gene Aliases
5830440B04, dy, mKIAA4087, mer, merosin
Chromosome Location
Chr. 10: 26981285 - 27617051 on Build GRCm38
UniGene ID
Mm.256087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008481.2 NP_032507.2 65 9398
GenBank mRNA AK076479.1 - 10 1414
AK164427.1 - 4639
AK220480.1 - 46 6604
BC025503.1 - 18 2473
BC039991.1 - 14 1871
BC054389.1 - 14 1877
U12147.1 - 65 9306

View Full Product Details