Gene Transcripts

Gene Symbol
Vmn2r14
Entrez Gene ID
231591
Gene Name
vomeronasal 2, receptor 14
Gene Aliases
EG231591
Chromosome Location
Chr. 5: 109215502 - 109224622 on Build GRCm38
UniGene ID
Mm.469882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104625.1 NP_001098095.1 2 372

View Full Product Details