Gene Transcripts

Gene Symbol
RS1
Entrez Gene ID
6247
Gene Name
retinoschisin 1
Gene Aliases
RS, XLRS1
Chromosome Location
Chr. X: 18639688 - 18672103 on Build GRCh38
UniGene ID
Hs.715725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000330.3 NP_000321.1 6 588
GenBank mRNA AF014459.1 - 6 568
AL049684.1 - 5 1044

View Full Product Details