Gene Transcripts

Gene Symbol
Ttc12
Entrez Gene ID
235330
Gene Name
tetratricopeptide repeat domain 12
Gene Aliases
E330017O07Rik
Chromosome Location
Chr. 9: 49436961 - 49486226 on Build GRCm38
UniGene ID
Mm.177413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172770.3 NP_766358.1 22 2161
GenBank mRNA AK054350.1 - 22 2162
BC056616.1 - 21 2089 4974294

View Full Product Details