Gene Transcripts

Gene Symbol
CCL1
Entrez Gene ID
6346
Gene Name
C-C motif chemokine ligand 1
Gene Aliases
I-309, P500, SCYA1, SISe, TCA3
Chromosome Location
Chr. 17: 34360328 - 34363233 on Build GRCh38
UniGene ID
Hs.72918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002981.2 NP_002972.1 1, 2 151
GenBank mRNA AK312183.1 - 1, 2 149
BC105073.1 - 1, 2 116 8144076
BC105075.1 - 1, 2 116 8144078
DA620119.1 - 1, 2 149
M57502.1 - 1, 2 129

View Full Product Details