Gene Transcripts
- Gene Symbol
- Gm8541
- Entrez Gene ID
- 667257
- Gene Name
- predicted gene 8541
- Gene Aliases
- EG667257
- Chromosome Location
- Chr. 2: 63525464 - 63575288 on Build GRCm38
- UniGene ID
- Mm.417007
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136239.1 |
- |
4 |
380 |
|
View Full Product Details